missing translation for 'onlineSavingsMsg'
Learn More

Camptothecin 97% MP Biomedicals

Product Code. 11465811
Click to view available options
Quantity:
25 mg
50 mg
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7689-03-4
Molecular Formula C20H16N2O4
Molecular Weight (g/mol) 348.36
MDL Number MFCD00081076
InChI Key VSJKWCGYPAHWDS-FQEVSTJZSA-N
Synonym camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecin
PubChem CID 24360
ChEBI CHEBI:27656
SMILES CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O

Specifications

Color Yellow
Density 1.51g/cm3
Assay Percent Range >97%
Quantity 250 mg
Formula Weight 348.3
Physical Form Solid
Chemical Name or Material Camptothecin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'