Learn More
Paragon Scientific™ Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids
For the determination of kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Brand:  Paragon Scientific™ CFO13NC
 View more versions of this product
Product Code. 15919989
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
10,000 to 50,000 cSt
45
ASTM D445, ISO 3104 and IP 71 Section 1
No
D 445, D 446
650
4 week lead-time