Learn More
Paragon Scientific™ Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids
For the determination of kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Brand: Paragon Scientific™ CFO09NC
This item is not returnable.
View return policy
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
240 to 1200 cSt | |
1.2 | |
ASTM D445, ISO 3104 and IP 71 Section 1 |
No | |
D 445, D 446 | |
400 |
Product Content Correction
Product Title
4 week lead-time
Spot an opportunity for improvement?Share a Content Correction