Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Click to view available options
Length (Metric):
10 mm
70 mm
75 mm
80 mm
90 mm
100 mm
120 mm
150 mm
Diameter (Metric) Outer:
1 mm
1.35 mm
1.55 mm
1.75 mm
2.35 mm
Unit Size:
Pack of 1000
Description
- Capillary tubes
- Available in a range of diameters/lengths
Specifications
Specifications
| Diameter (Metric) Inner | 1.15 mm |
| Diameter (Metric) Outer | 1.55 mm |
| For Use With (Application) | Melting point determination |
| Tube Style | One end closed |
| Length (Metric) | 75 mm |
| Type | Capillary Tube |