Learn More
Erlab™ Captair by Erlab™ Stainless Steel Work Surfaces
Product Code. p-8011761
Click to view available options
For Use With (Equipment):
CaptairFlow 391 Ductless Fume Hood
CaptairFlow 483 Ductless Fume Hood
CaptairFlow 714 Ductless Fume Hood
Unit Size:
Each
Product Code. 13416583
Brand: Erlab™ I0116050001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Stainless Steel Work Surfaces are for use with Captair PCR work stations.
  • High chemical and mechanical resistance
  • Rounded corners to facilitate cleaning operations
  • Built in spill tray
Specifications
Material Stainless Steel
For Use With (Equipment) CaptairFlow 391 Ductless Fume Hood