Learn More
Erlab™ Captair Smart Trespa™ Work Surface
Product Code. p-7160209
Click to view available options
For Use With (Equipment):
Smart 321 Fume Hood
Smart 391 Fume Hood
Smart 392 Fume Hood
Smart 481 Fume Hood
Smart 483 Fume Hood
Smart 633 Fume Hood
Smart 714 Fume Hood
Unit Size:
Each
Product Code. 15545154
Brand: Erlab™ I0160030001
Erlab™ Captair Smart Trespa™ Work Surface is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
  • Made of phenolic resin with ergonomic arm for a comfortable working position
  • High chemical and mechanical resistance
  • Ideal for precision weighing operations
Specifications
For Use With (Equipment) Smart 321 Fume Hood
Includes Spill Tray
Depth (Metric) 510 mm
Width (Metric) 694 mm
Product Type Work Surface
Material Phenolic Resin