missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ MMC ImpRes Multimodal BioProcess™ Chromatography Medium

Product Code. p-4625999 Shop All Cytiva Products
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 15131556

Brand: Cytiva 17371601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules

  • High yields achieved through high-resolution beads and ligand selectivity
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • Enables use of platform approach to MAb process development
  • Displays broad pH/salt operational window

Avoid using oxidizing agents, cationic detergents.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
Stability 1 M Acetic Acid, 1 M NaOH, 2 M NaCl, 5% 1-Propanol, 30% Isopropanol, 70% Ethanol
For Use With (Application) For High Resolution Polishing of Monoclonal Antibodies (MAbs) and Other Biomolecules
Storage Requirements 4°C to 30°C
Anion Exchange Capacity 25 to 39 μM/mL
Binding Capacity 60 to 90 mg Mab/mL
Product Type Capto MMC ImpRes Media
Particle Size 40 μm
Max. Pressure 300 kPa
Matrix High-flow Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Multimodal Weak CatIon Exchanger
Show More Show Less
compliance-icons
Product Identifier
  • Capto MMC ImpRes, 25 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.