missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon, Hanna Instruments™
Click to view available options
Quantity:
50 Pc.
Specifications
Specifications
CAS | 7440-44-0 |
Molecular Formula | C |
Quantity | 50 Pc. |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Molecular Weight (g/mol) | 12.01 |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'