Learn More
Swann-Morton™ Carbon Steel Scalpel Blade Spare
Replacement blades for knife set 12942545
7.85 EUR - 11.85 EUR
Specifications
Blade Material | Carbon Steel |
---|---|
For Use With (Equipment) | SCA-500-K |
Thickness (Metric) | 0.4 mm |
Quantity | 50/Pk. |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11330905
|
Swann-Morton™
1242 |
# 2 |
9.00 EUR
Pack of 50 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
11320905
|
Swann-Morton™
1241 |
# 1 |
7.85 EUR
Pack of 50 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
11340905
|
Swann-Morton™
1243 |
# 3 |
11.85 EUR
Pack of 50 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
Slotted plastic handle with clamping screw to grip blade. With blades Nos. 1 and 2.Specifications
Carbon Steel | |
SCA-500-K | |
0.4 mm | |
50/Pk. |