missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A2/CPA2 Antibody (OTI1C7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-01992
This item is not returnable.
View return policy
Description
Carboxypeptidase A2/CPA2 Monoclonal specifically detects Carboxypeptidase A2/CPA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Carboxypeptidase A2/CPA2 | |
Monoclonal | |
0.89 mg/mL | |
Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
CPA2 | |
Full length human recombinant protein of human CPA2(NP_001860) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
1358 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI1C7 | |
Unconjugated | |
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), EC 3.4.17, EC 3.4.17.15 | |
Mouse | |
44.9 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction