Learn More
Fisherbrand™ Cardboard Cryobox Inserts, 133mm
Product Code. p-8013110 Shop All Fisherbrand Products
Click to view available options
Height (Metric):
25mm
30mm
40mm
65mm
Array:
10 x 10
16 x 16
4 x 4
7 x 7
8 x 8
9 x 9
Unit Size:
Pack of 10
Product Code. 11842513
Only null left
Add to basket
Add to basket
Combine these inserts with Cryoboxes for flexible cell storage and sample handling. Fisherbrand™ Cardboard Cryobox Inserts feature ultra-low temperature resistant cardboard with water repellent finish for sample storage.
  • Ultra low temperature-resistant cardboard
  • For sample storage
  • Temperature-resistant down to -80°C
  • Water-repellent finish
Specifications
Quantity 10 Pack
Material Cardboard
Product Type Cryo Box Insert
Height (Metric) 25mm
Width (Metric) 133mm
Length (Metric) 133mm
Array 10 x 10