Learn More
Zeiss™ Light Guide Holder for Stemi™ Mounts
Product Code. 11894734
Click to view available options
For Use With (Equipment):
Stemi Mounts
Unit Size:
Each
Product Code. 11894734
Brand: Zeiss™ 4170859002000
Only null left
Add to basket
Add to basket
Designed for use with Zeiss Microscopes
Compatible Microscopes:
  • Stemi 2000
  • Stemi 2000 C
  • Stemi 2000 CS
  • Stemi 305
  • Stemi 305 Demo
  • Stemi 305 trino
  • STEMI SV 11
  • Stemi SV 11 Apo
  • STEMI SV 6
  • SteREO Discovery.V12
  • SteREO Discovery.V8
Specifications
Type Light Guide Holder
For Use With (Equipment) Stemi Mounts
Includes Light guide holder