Learn More
Motic™ CCIS™ Plan Achromatic Phase Objective
Click to view available options
Type:
PL Ph0 4X
PL Ph1 10X
Focus Distance:
0.1 mm, WD=12.6 mm
0.25 mm, WD=4.1 mm
Unit Size:
Each
Description
- For use with AE31 Inverted Microscope
- Achromatic Plan Phase Objective
- Flat field
- Eliminates color aberration
- Magnification and Numerical Apertures available:
- 10X/0.25 (WD=4.1mm) Ph1 for AE
- 4X/0.10 (WD=12.6mm) Ph0 for AE
Specifications
Specifications
| Type | PL Ph1 10X |
| Focus Distance | 0.25 mm, WD=4.1 mm |
| For Use With (Equipment) | AE31 inverted microscope |
| Quantity | 1 Piece |