Learn More
Fisherbrand™ Catalyst PharMed™ BPT Formulation Stopped Tubing Assembly
Product Code. p-6877023 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
0.8 mm
1.6 mm
3.2 mm
4.8 mm
6.4 mm
8 mm
9.5 mm
Thickness (Metric):
1.6 mm
2.4 mm
Unit Size:
Pack of 12
Product Code. 15531325
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Preconfigured ready-to-use links and assemblies.
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Ensures optimal amount of tubing is in pump head
Specifications
Description Stopped Tubing Assembly
Diameter (Metric) Inner 0.8 mm
Thickness (Metric) 1.6 mm