Learn More
Invitrogen™ CD137 Ligand (4-1BB Ligand) Monoclonal Antibody (5G11), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 14-9056-82
Description
This 5G11 monoclonal antibody recognizes human CD137 ligand also known as 4-1BB ligand, TNFSF9, or CDw137L. CD137 ligand is a member of the TNF superfamily, and is a type II membrane protein. It is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation of these cells. Its receptor, CD137 (4-1BB) is found on a variety of cells, including inflamed endothelial cells, where it enhances extravasation of CD137 ligand-expressing monocytes. Activated T cells also express CD137, and engagement with CD137 ligand enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137 ligand is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but not on Hodgkin lymphoma and T-cell lymphoma). This 5G11 antibody has been tested by immunohistochemical staining of formalin-fixed paraffin embedded human tissue using low pH antigen retreival and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
CD137 ligand (CD137L) is a type II membrane protein and part of the TNF superfamily. CD137L is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation. Its receptor, CD137/4-1BB, is found on a variety of cells, including inflamed endothelial cells, where its expression enhances extravasation of CD137L expressing monocytes. Activated T cells also express CD137, and engagement with CD137L enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137L is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but it is lacking on Hodgkin lymphoma and T-cell lymphoma). CD137L is also expressed in carcinoma cell lines, and is thought to be involved in T cell-tumor cell interactions.
Specifications
| CD137 Ligand (4-1BB Ligand) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P41273 | |
| TNFSF9 | |
| Affinity chromatography | |
| RUO | |
| 8744 | |
| 4°C, do not freeze | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 5G11 | |
| Unconjugated | |
| TNFSF9 | |
| 4 1 BB Ligand; 4 1 BBL; 41BB Ligand; 4-1BB ligand; 41BBL; 4-1BBL; 4-1BB-L; AI848817; CD 137 Ligand; CD 137L; CD137 Ligand; CD137L; Cd157l; CDw137L; costimulatory molecule; homolog of mouse 4-1BB-L; Ly63l; receptor 4-1BB ligand; TNFRSF9; Tnfsf9; TNLG5A; tumor necrosis factor (ligand) superfamily member 9; tumor necrosis factor (ligand) superfamily, member 9; tumor necrosis factor ligand 5A; tumor necrosis factor ligand superfamily member 9; tumor necrosis factor superfamily member 9 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only