missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Antibody (M5E2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 11 publications
191.00 EUR - 343.00 EUR
Specifications
| Antigen | CD14 |
|---|---|
| Clone | M5E2 |
| Concentration | 1.0 mg/mL |
| Dilution | Simple Western 50 ug/ml, Flow Cytometry < 2.0 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready, Block/Neutralize reported in scientific literature (PMID 27228163), Dual RNAscope ISH-IHC |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18250005
|
Novus Biologicals
NB100-77758SS |
0.025 mg |
191.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18725133
|
Novus Biologicals
NB100-77758 |
0.1 mg |
343.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD14 Monoclonal specifically detects CD14 in Human, Porcine, Bovine, Canine, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready, Block/Neutralize, Dual RNAscope ISH-IHC.Specifications
| CD14 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Asthma, Cancer, Cell Biology, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| CD14 | |
| IgG2a κ | |
| Protein G purified |
| M5E2 | |
| Simple Western 50 ug/ml, Flow Cytometry < 2.0 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready, Block/Neutralize reported in scientific literature (PMID 27228163), Dual RNAscope ISH-IHC | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Pig, Bovine, Canine, Primate | |
| 929 | |
| Human peripheral blood cells. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title