missing translation for 'onlineSavingsMsg'
Learn More

CD152 (CTLA-4) Monoclonal Antibody (14D3), PE-eFluor™ 610, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

181.00 EUR - 418.00 EUR

Specifications

Antigen CD152 (CTLA-4)
Clone 14D3
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16361084
View Documents
Invitrogen
61-1529-41
25 Tests
181.00 EUR
Pack of 25
Please to purchase this item. Need a web account? Register with us today!
16371084
View Documents
Invitrogen
61-1529-42
100 Tests
418.00 EUR
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mouse Monoclonal Antibody

CTLA4 (Cytotoxic T-Lymphocyte Antigen 4) is a CD28-family receptor expressed on mainly CD4+ T cells. It binds the same ligands as CD28 (CD80 and CD86 on B cells and dendritic cells), but with higher affinity than CD28. However, in contrast to CD28 which enhances cell function when bound at the same time as the T cell receptor, CTLA4 inhbits the T cell and prevents it from functioning. Intracellular CTLA4 is also found in regulatory T cells and may be important to their function.
Specifications

Specifications

CD152 (CTLA-4)
5 μL/Test
Monoclonal
Liquid
RUO
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
ALPS5; CD; CD152; CD152 antigen; CD152 isoform; CD152 protein; CD152 protein precursor; celiac disease 3; CELIAC3; Ctla4; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; cytotoxic T-lymphocyte associated protein 4; cytotoxic T-lymphocyte protein 4; cytotoxic T-lymphocyte protein 4 isoform CTLA4-TM; cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; EGK_04712; GRD4; GSE; ICOS; IDDM12; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; Ly-56; sCTLA4; soluble form; transmembrane form
CTLA4
Primary
4° C, store in dark, DO NOT FREEZE!
CTLA4
14D3
Flow Cytometry
PE-eFluor 610
Mouse
Human, Rhesus Monkey
P16410
1493, 705673
IgG2a κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications