Learn More
CD1d Mouse, anti-Human, CoraLite Plus 488, Clone: 1C12F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66257-100UL
This item is not returnable.
View return policy
Description
The CD1d antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD1d. This antibody recognizes human antigen. The CD1d antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| CD1d | |
| 1C12F1 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG1 |
| Western Blot | |
| 1000 μg/mL | |
| CD1A, CD1D, CD1d molecule, R3, R3G1 | |
| 100 μL | |
| 912 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido