missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD22 / BL-CAM Mouse, Clone: BLCAM/1795, enQuireBio™
Mouse Monoclonal Antibody
199.00 EUR - 458.00 EUR
Specifications
Antigen | CD22 / BL-CAM |
---|---|
Clone | BLCAM/1795 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951198
|
EnquireBio
933-MSM5-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15961198
|
EnquireBio
933-MSM5-P1 |
100 μg |
458.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD22 / BL-CAM | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
933 | |
Recombinant human BL-CAM/CD22 protein (aa52-178) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
130-140kDa |
BLCAM/1795 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001172030 | |
CD22 | |
IgG1 κ | |
Protein A/G Purified | |
Cell Surface | |
Raji, Daudi, IM9, JY25 and human peripheral blood lymphocytes. Human Tonsil |