missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, Clone: B-B11, Novus Biologicals™

Mouse Monoclonal Antibody

393.08 EUR - 745.12 EUR

Specifications

Antigen CD3
Clone B-B11
Concentration 1 mg/mL
Dilution Flow Cytometry, Functional
Applications Flow Cytometry, Functional Assay
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18320316
View Documents Promotion Details
Bio-Techne
NBP3-14571-200UL
200 μg
393.08 EUR
200µL
Please to purchase this item. Need a web account? Register with us today!
18331515
View Documents Promotion Details
Bio-Techne
NBP3-14571-500UL
500 μg
745.12 EUR
500µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry, Functional Assay
Specifications

Specifications

CD3
1 mg/mL
Flow Cytometry, Functional Assay
Unconjugated
Mouse
Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells
PBS. Sterile-filtered through 0.22 um.
916
IgG1 κ
Affinity purified
Antibody
B-B11
Flow Cytometry, Functional
Monoclonal
Purified
RUO
Human
CD_antigen: CD3e, CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, IMD18, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
PHA activated T cells
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications