Learn More
CD68 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25747-1-AP-20UL
 View more versions of this product
Product Code. 16804315
  • 140.00 EUR / 20µL
Explore more special offers
Description
Description
These complexes include CRSP (for cofactor required for Sp1 activation), which is required, in conjunction with TAFIIs, for transcriptional activation by Sp1. CRSP is ubiquitously expressed in various tissues and functions as a multimeric complex that consists of nine distinct subunits. Several members of the CRSP family share sequence similarity with multiple components of the yeast transcriptional mediator proteins, including CRSP150, which is related to yeast Rgr1, and CRSP70, which is similar to the elongation factor TFIIS. CRSP77 and CRSP150 are also related to proteins within the putative murine mediator complex, while CRSP130 and CRSP34 are largely unrelated to either murine or yeast proteins.
Specifications
Specifications
CD68
Polyclonal
Unconjugated
CD68
CD68
CD68 Fusion Protein Ag22815
20 μL
Primary
Human, Mouse
Antibody
IgG
Flow Cytometry, Immunoprecipitation
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P31996, P34810
Rabbit
Antigen Affinity Chromatography
RUO
12514, 968
-20°C
Liquid