missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC25A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55031-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to CDC25A.
Replication factor C (RFC) is an essential DNA polymerase accessory protein that is required for numerous aspects of DNA metabolism, including DNA replication, DNA repair and telomere metabolism. RFC is a heteropentameric complex that recognizes a primer on a template DNA, binds to a primer terminus and loads proliferating cell nuclear antigen (PCNA) onto DNA at primer-template junctions in an ATP-dependent reaction. All five of the RFC subunits share a set of related sequences (RFC boxes) that include nucleotidebinding consensus sequences. Four of the five RFC genes (RFC1, RFC2, RFC3 and RFC4) have consensus ATP-binding motifs. The small RFC proteins, RFC2, RFC3, RFC4 and RFC5, interact with Rad24, whereas the RFC1 subunit does not. RFC2, the third-largest subunit of the RFC complex, exhibits ATP binding which makes it important for both DNA replication and checkpoint function.Specifications
CDC25A | |
Polyclonal | |
Unconjugated | |
CDC25A | |
CDC25A, CDC25A2, M phase inducer phosphatase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12530, 993 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.53 mg/mL | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P30304, P48964 | |
Cdc25a | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |