Learn More
CDCA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513564
This item is not returnable.
View return policy
Description
F-box-like protein which is required for entry into mitosis. Acts by participating in E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase. [UniProt]Specifications
CDCA3 | |
Polyclonal | |
Unconjugated | |
CDCA3 | |
2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
14793, 83461 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1.8 mg/mL | |
PBS with 0.09% sodium azide | |
Q99618, Q99M54 | |
CDCA3 | |
KLH conjugated synthetic peptide between 41-69 amino acids from the N-terminal region of human CDCA3 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |