Learn More
Lonza™ Cell Line Nucleofector™ Kit R
Brand:  Lonza™ VCA-1001
Product Code. 11566329
  • 1015.00 EUR / 25 reactions
Explore more special offers
Specifications
99% Transfection efficiency with siRNA duplexes even in suspension cells.
Five different Cell Line Nucleofector™ Solutions: C, L, R, T, and V
Cell Line Nucleofector Kit R
Nucleofector™ Device
25
1 Ea.