Learn More
Corning™ Cell Recovery Solution
GREENER_CHOICE
Product Code. p-8874000 Shop All Corning Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 11543560
Brand: Corning™ 354253
Allows for the recovery of cells cultured on Corning™ Matrigel™ Basement Membrane Matrix for subsequent biochemical analyses
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Product Type Cell recovery solution
Quantity 100 mL