Learn More
Greiner Bio-One CELLSTAR TC treated Cell Culture Dish with vents
Click to view available options
Volume (Metric) Working:
16 to 17 mL
25 to 27 mL
3 mL
6 to 7 mL
Unit Size:
Pack of 120
Pack of 360
Pack of 600
Pack of 740
Description
- High-quality virgin polystyrene construction
- Nonpyrogenic
- Free of RNase and DNase
- Stackable
- Uniform gas exchange and reduced condensation
Applications
- For optimal cell attachm
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Specifications
| Closure Color | Clear |
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Capacity (Metric) | 28 mL |
| Color | Clear |
| Culture Area | 21 sq. cm |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Sterile |
| Show More |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Greiner Bio-One CELLSTAR TC treated Cell Culture Dish with vents >
Spot an opportunity for improvement?Share a Content Correction