Learn More
Greiner Bio-One CELLSTAR TC treated Cell Culture Dish with vents
Provides uniform gas exchange and reduced condensation.
Brand: Greiner Bio-One 639160
This item is not returnable.
View return policy
Description
- High-quality virgin polystyrene construction
- Nonpyrogenic
- Free of RNase and DNase
- Stackable
- Uniform gas exchange and reduced condensation
Applications
- For optimal cell attachm
Specifications
Clear | |
Polystyrene | |
240 mL | |
143 sq. cm | |
Round | |
Physical Surface Treatment | |
25 to 27 mL | |
120 | |
20 mm |
Closure Included | |
Flat | |
Clear | |
Polystyrene | |
Sterile | |
Cell Culture Dish | |
Lid | |
5 | |
145 mm |