Learn More
Biodesign™ Cellulose Dialysis Tubing Strips
Product Code. p-8009621
Click to view available options
MWCO:
14 kDa
3.5 kDa
8 kDa
Capacity (Metric):
0.32 mL/cm
1.32 mL/cm
1.91 mL/cm
19.13 mL/cm
3.57 mL/cm
6.45 mL/cm
Unit Size:
Pack of 50
Product Code. 12727476
Brand: Biodesign™ D011
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.
  • Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
  • Just wet in distilled water and use
  • Available in 305mm strips
  • Specially purified for life science research
  • Supplied in an airtight, moisture-proof container for maximum shelf life
  • Available in 14000Da, 8000Da, and 3500Da molecular weight cut offs
Specifications
Product Type Dialysis Tubing Strips
MWCO 14 kDa
Material Cellulose
Diameter (Metric) 6.4 mm
Length (Metric) 30.5 m
Capacity (Metric) 1.32 mL/cm
Color Transparent
Quantity 1 Each
For Use With (Application) For life sciences research