Learn More
MP Biomedicals™ CellVation™ Cryopreservation Medium
Recommended for the preservation of stem cells, hepatocytes, tissue samples, and a wide variety of cell types and established cell lines
Brand: MP Biomedicals™ 0920300M2
Description
CellVation is a cryopreservation medium designed for optimal cell recovery and convenience. It does not contain DMSO or serum, and is scientifically engineered for serum-free cultures, as well as, ideally suited for all types of serum. It has also been shown to preserve the potential of stem cells for differentiation.

Specifications
Solution | |
Cell counting, viability and cryopreservation | |
Mycoplasma, Bacteria, Fungi, Yeast and Mold free. Certified U.S. origin. | |
6.5 to 7.5 | |
CellVation Cryopreservation Media |
Cryopreservation Media | |
60 mL | |
<0.5 EU/mL | |
Store at 4°C |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.