Learn More
Kimberly-Clark™ Centrefeed Roll Wiper Dispenser
Product Code. 10212381 Shop All Kimberly-Clark Professional Products
Click to view available options
Color:
Gray
White
Unit Size:
Each
Product Code. 10212381
Brand: Kimberly-Clark™ 7088
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for maintaining hygiene in busy food areas. Centrefeed Roll Wiper Dispenser is compatible with a large range of wipers, hand towels, and complies with food safety standards.
  • Dispenses sheets of wiper roll without the need to touch the dispenser, helping to reduce contamination and the spread of germs
  • A smart, white, water-resistant wiper dispenser that is ideal for maintaining hygiene in laboratory, healthcare and general work areas
  • Wall-mountable, ideal for cleaning stations
  • Easy to refill, keep clean and maintain
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wiper Dispenser
Color White
Depth (English) 10.8 in.
Depth (Metric) 27.4 cm
Width (English) 10.5 in.
Width (Metric) 26.6 cm
Height (English) 15.2 in.
Height (Metric) 38.7 cm
Holds Wipes
Material Plastic
Mounting Wall or Surface Mount
No. of Compartments 1
Quantity 1 Ea.