Learn More
Eppendorf Centrifuge 5427 R (EU-IVD)
BIOPHARMAGREENER_CHOICE
Product Code. 17834754 Shop All Eppendorf Products
Click to view available options
Electrical Requirements:
230V, 50/60Hz (EU)
Includes:
With Rotor FA-45-30-11
With Rotor FA-45-48-11
Without rotor
Unit Size:
Each
Product Code. 17834754
Brand: Eppendorf 5429000315
Centrifuge 5427 R contains a natural refrigerant (R290) to protect your samples and the environment. It features 9 different rotor options including an aerosol-tight 48-place rotor for high-throughput applications.
  • Rotor capacity of 48 x 1.5/2.0 mL and 12 x 5.0 mL
  • QuickLock rotor for easy lid locking
  • Max. speed of 25,001 x g (16,220 rpm)
  • High-capacity 48-place rotor increases productivity and saves time
  • Soft-touch one-finger closure for ergonomic operation
  • Temperature range from -11 °C to 40 °C
  • FastTemp function for quick pre-cooling (11 minutes from ∼23°C to 4°C)
  • ECO shut-off programmable after 1, 2, 4 or 8 hours of non-use for energy savings and compressor life
  • Built-in condensation drain eliminates water accumulation
  • Automatic rotor recognition and imbalance detection for safety
TRUSTED_SUSTAINABILITY
Specifications
Capacity 48 x 1.5/2.0 mL, 12 x 5.0 mL
Max. Speed 16,220 rpm
Max. RCF 25,001 x g
Display Large, brightly lit LCD
Height (English) Exterior 10 in.
Height (Metric) Exterior 25.4 cm
Noise Level < 55 dBA with rotor FA-45-48-11
Timer 5 sec. to 9:59 hr.
Width (English) Exterior 12.6 in.
Width (Metric) Exterior 31.9 cm
Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
Type Microcentrifuge
Includes With Rotor FA-45-30-11
Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, certified aerosol-tight rotor lids
Electrical Requirements 230V, 50/60Hz (EU)
Model Centrifuge 5427 R, rotary knobs
Wattage 495 W
Refrigerated Yes
Voltage 230 V
Show More Show Less
The Centrifuge 5427 R is a non-automatic centrifuge for the separation of liquid substance mixtures from the human body and is specifically intended to be used as an accessory together with an in vitro diagnostic device and to enable its use according to its intended purpose. This product complies with the requirements of the EU In Vitro Diagnostics Regulation (IVDR) 2017/746. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.