Learn More
Gestigkeit Harry CERAN™ Hotplate with Separate Wall Mounted Regulator
CERAN™ Hotplates are highly resistant to breakage and temperature changes, will not distort, permeable to ultra-violet light and also highly acid-resistant.
Brand:  Gestigkeit Harry 4SR
 View more versions of this product
Product Code. 11764838
  • 1360.00 EUR / Each
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Description
  • CERAN™ is an extraordinary glass ceramic - highly resistant to breakage and temperature changes, will not distort, permeable to ultra-violet light and also highly acid-resistant.
  • Construction: the low-mass CERAN™ plate is electrically heated over its full surface, and is embedded into a high-grade, stainless steel frame which is mounted on an internally insulated stainless steel housing.
  • The temperature control system is regulated by an electronic NiCr-Ni temperature sensor (regulation range between 50°C and 500°C). The controller is built into a pressure-cast aluminum casing. The heating time for 500°C is approximately 8 minutes.
  • Four feet, which can be individually adjusted to an approximate height of 8 mm, ensure a wobble-free, horizontal stability.
  • Type 4 SR is also equipped with 4 retort rod sockets (situated 100 mm from each other). In the EB Series, the electrical connection between the controller and the hot plate is protected by a flexible metal hose.
Specifications
Specifications
Stainless Steel
Hotplate
Ceramic
5 kg
50/60 Hz
180 x 440 x 100 mm
230 V
50°C to 500°C
1500 W
4 SR
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products