missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGP 12177 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Partial agonist at β3-adrenoceptors. β1- and β2-adrenoceptor antagonist (Ki values are 0.9, 4 and 88 nM for β1, β2 and β3 receptors respectively).
Specifications
Specifications
Quantity | 10 mg |
Synonym | cgp 12177 hydrochloride, 4-3-tert-butylamino-2-hydroxypropoxy-1h-benzo d imidazol-2 3h-one hydrochloride, +/--cgp-12177a hydrochloride, cgp 12177 hydochloride, y-cgp-12177a hydrochloride, ?-cgp-12177a hydrochloride, inverted exclamation marka-cgp-12177a, 1,3-dihydro-2h-benzimidazol-2-one hydrochloride, inverted question mark-cgp-12177a hydrochloride, +/--cgp-12177a hplc , solid |
Formula Weight | 315.8 |
Percent Purity | >98% |
Chemical Name or Material | CGP 12177 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction