Learn More
Thermo Scientific™ Chest Freezer Racks
Alle ansehen Thermo Scientific Produkte
missing translation for 'orderingAttributeHoverText'
Holds:
Boxes (2 in.)
Boxes (3 in.)
General Freezer Items
Plates
For Use With (Equipment):
12.7 cu. ft. Freezers
12.7, 17, 20 cu. ft. Freezers
12.7, 20 cu. ft. Freezers
13, 17, 20 cu. ft. Freezers
20 cu. ft. Freezers
3 cu. ft. Chest Freezers
3 cu. ft. Freezers
missing translation for 'unitSize'
Each
Beschreibung
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
- Warranty varies by country, please contact us for details
Spezifikation
Spezifikation
| Product Type | Microplate Rack |
| Color | Silver |
| Holds | Plates |
| Positions | 27 |
| Shape | Rectangular |
| For Use With (Equipment) | 12.7, 17, 20 cu. ft. Freezers |
| Freezer Compatibility (Capacity) | 12.7, 17, 20 cu. ft. |
| Width (Metric) | 13.5 cm |
| Width (English) | 5.3 in. |
| Height (Metric) | 65.5 cm |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur