Learn More
WTW™ Chlorine Dioxide Test Kits
Brand: WTW™ 252017
This item is not returnable.
View return policy
Description
Specifications
Colorless | |
For drinking water, waste water, disinfectants | |
Certificates | |
2°C to 8°C | |
CombiCheck Manganese (Mn) Reagent Test Kit |
5 mL | |
For PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers | |
Manganese | |
Formaldoxime |