missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, ACS Reagent Grade, Stabilized with Ethanol, Scharlab
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
- Synonyms: Trichloromethane, Formyl trichloride
- Molecular Formula: CHCl3
- Molar Mass: 119.38g/mol
- CAS: 67-66-3
- EIN ECS No.: 200-663-8
- Dielectric Constant: 4.8 (20°C)
- Suitability for Use in Dithizone Tests: Passes test
Specifications
Specifications
| Melting Point | -63°C |
| CAS | 67-66-3 |
| Appearance | Clear |
| Density | 1.47g/cm3 |
| Boiling Point | 61°C |
| Quantity | 1 L |
| Identification | Passes Test (IR spectrum) |
| Ignition Point | 982°C |
| Physical Form | Liquid |
| Residue after Evaporation | 0.0005% max |
| Show More |
Safety and Handling
Product Identifier
- Chloroform, reagent grade, ACS, ISO, stabilized with ethanol
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction