Learn More
Thermo Scientific Chemicals Chlorophenol Red, sodium salt
59.40 EUR - 197.00 EUR
Chemical Identifiers
CAS | 123333-64-2 |
---|---|
Molecular Formula | C19H11Cl2O5S |
Molecular Weight (g/mol) | 422.25 |
MDL Number | MFCD00151199 |
InChI Key | QXTPRQZMDKBTAI-UNOMPAQXSA-M |
Synonym | 3', 3''-Dichlorophenolsulfonephthalein, sodium salt |
PubChem CID | 131846007 |
SMILES | OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10092840
|
Thermo Scientific Chemicals
216310100 |
10 g | Glass bottle |
59.40 EUR
10g |
In Stock
Log in to see stock. |
||||
11976811
|
Thermo Scientific Chemicals
216310500 |
50 g | Glass bottle |
197.00 EUR
50g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123333-64-2 | |
422.25 | |
QXTPRQZMDKBTAI-UNOMPAQXSA-M | |
131846007 |
C19H11Cl2O5S | |
MFCD00151199 | |
3', 3''-Dichlorophenolsulfonephthalein, sodium salt | |
OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O |
Specifications
267.0°C | |
C19H11Cl2O5S | |
3', 3''-Dichlorophenolsulfonephthalein, sodium salt | |
QXTPRQZMDKBTAI-UNOMPAQXSA-M | |
OC1=CC=C(C=C1Cl)C(=C1/C=CC(=O)C(Cl)=C1)\C1=CC=CC=C1S([O-])(=O)=O | |
422.25 | |
445.25 | |
(575nm, 0.1N NaOH) ca. 1200 | |
Brown | |
Chlorophenol Red, sodium salt |
123333-64-2 | |
MFCD00151199 | |
Solubility in water: soluble. | |
Authentic | |
2-chloro-4-[3-(3-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol;sodium | |
131846007 | |
Pure | |
From pH 4.6 (yellow) to pH 7.0 (purple) | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
- Chlorophenol Red, sodium salt, water soluble
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only