Learn More
Tocris Bioscience™ CHR 6494 trifluoroacetate
Potent and selective haspin inhibitor
Brand:  Tocris Bioscience™ 5326/10
178.83 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18780278
  • 202.00 EUR / 10mg
Explore more special offers
Description
Description
CHR 6494 trifluoroacetate is a potent and selective haspin kinase inhibitor (IC50 = 2 nM). Selective over 27 other protein kinases including Aurora B kinase. Induces cell cycle arrest at G2/M, inhibits cell proliferation and promotes apoptosis in multiple cancer cell lines. Inhibits angiogenesis ex vivo and attenuates tumor growth of HCT-116 xenografts in rats.
Specifications
Specifications
>98%
CHR 6494 trifluoroacetate
CHR 6494 trifluoroacetate
10 mg
406.36