Learn More
Macherey-Nagel™ Chromabond™ Florisil Polypropylene SPE Column
Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
200 mg
500 mg
1 g
2 g
Column Capacity (Metric):
3 mL
6 mL
Unit Size:
Each
Pack of 30
Pack of 50
Specifications
Specifications
| Base Material | Magnesium Silicate |
| Column Capacity (Metric) | 6 mL |
| Column Type | SPE |
| Bed Weight | 500 mg |
| Particle Size | 150 to 250 μm |
| Sorbent | Chromabond Florisil |
| Type | Solid Phase Extraction Column |