Learn More
Macherey-Nagel™ Chromabond™ Glass Empty SPE Column
Shop All Macherey-Nagel Products
Click to view available options
Column Capacity (Metric):
3 mL
6 mL
Unit Size:
Pack of 30
Pack of 50
Specifications
Specifications
| Column Capacity (Metric) | 6 mL |
| Column Type | Empty |
| For Use With (Application) | Solid Phase Extraction |
| Type | Empty Solid Phase Extraction Column |
| Includes | Glass fiber frits, filter element |