Learn More
Lenz Laborglasintrumente Chromatographic Columns with Indentations and NS Socket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05440708
Product Code. 10375831
  • 114.00 EUR / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Description
  • Basic chromatographic columns
  • With indentations above the stopcock for placing a cotton plug
  • With NS socket
  • With PTFE stopcock and retaining device
  • Made of DURAN™ tubing
Specifications
Specifications
1 L
NS 29/32
Duran Glass
40 mm