missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chrome Azurol S, MP Biomedicals™
Click to view available options
Quantity:
10 g
25 g
100 g
Specifications
Specifications
Formula Weight | 605.29 |
Color | Brown |
Quantity | 100 g |
Physical Form | Powder |
Chemical Name or Material | Chrome Azurol S, Dye content: approx. 65% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction