missing translation for 'onlineSavingsMsg'
Learn More

Chrome Azurol S, MP Biomedicals™

Product Code. 11427554
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chrome Azurol S is used as a reagent for determination of fluoride

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1667-99-8
Molecular Formula C23H13Cl2Na3O9S
Molecular Weight (g/mol) 605.277
MDL Number MFCD00001615
InChI Key FUIZKNBTOOKONL-UHFFFAOYSA-K
Synonym Mordant Blue 29, C.I. 43825
PubChem CID 54736314
IUPAC Name trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate
SMILES CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]

Specifications

Formula Weight 605.29
Color Brown
Quantity 10 g
Physical Form Powder
Chemical Name or Material Chrome Azurol S, Dye content: approx. 65%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.