missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium for AAS 10g/L HCl matrix, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Packaging | Bottle |
| CAS | 7697-37-2 |
| Physical Form | Liquid |
| Molecular Formula | HNO3 |
| Concentration | 10 g/L ±0.02 g/L |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| Type | Single-element Concentrated Standard Solution |
| SMILES | O[N+]([O-])=O |
| IUPAC Name | nitric acid |
| Show More |
Safety and Handling
Product Identifier
- Single-element Standard Solution for AAS. Chromium (Cr) 10000mg/l in HCl 5%
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction