Learn More
Proteintech ChromoTek Myc-Trap™ Magnetic Agarose
The ChromoTek Myc-Trap™ Magnetic Agarose is an affinity resin for IP of Myc-fusion proteins.
Brand: Proteintech ytma-400
This item is not returnable.
View return policy
Description
- Consists of a Myc Nanobody/VHH coupled to magnetic agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
Myc-tag sequence motif EQKLISEEDL at the N-terminus, C-terminus, or internal site of the fusion protein. Endogenous c-myc is NOT bound. | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose | |
400 Reactions |