missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ciclopirox

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ciclopirox is a pan-histone demethylase inhibitor. Exhibits no significant effect on histone deacetylases. Inhibits MYC-signaling and proliferation of neuroblastoma cells. Promotes Cdc25A-degradation in cancer cells. Antifungal.
Specifications
Specifications
| Chemical Name or Material | 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone |
| CAS | 29342-05-0 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Molecular Formula | C12H17NO2 |
| Formula Weight | Observed MW: 207.27 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction