Learn More
Thermo Scientific™ Cimarec™ i Benchtop Distributors (Link and Sync)
Ensure identical operating conditions for multiple stirrer installations by using these benchtop distributors, for use with Mini, Micro, Compact and Maxi series stirrers.
Brand:  Thermo Scientific™ 50091721
 View more versions of this product
Product Code. 10247081
  • 355.77 EUR / Each
Estimated Shipment: 31-01-2025
Description
Ensure identical operating conditions for multiple stirrer installations by using Thermo Scientific™ Cimarec™ i Benchtop Distributors (Link and Sync), for use with Mini, Micro, Compact and Maxi series stirrers.
  • Simultaneous control of 2, 4, 6, or 8 Mini, Micro or Compact series stirrers using a single Telemodul 20 C or Telemodul 40 C external controller
  • Simultaneous control of 2 or 4 Maxi Series stirrers using a single Telemodul 20 C or Telemodul 40 C external controller
  • Even-powered distribution for synchronized stirring
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8-Place Distributor
Mini, Micro, Compact, and Maxi Series Stirrer
Benchtop Distributor