missing translation for 'onlineSavingsMsg'
Learn More

Cinnamic Aldehyde, 99%, MP Biomedicals

Product Code. 11462052 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 104-55-2
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
InChI Key KJPRLNWUNMBNBZ-QPJJXVBHSA-N
Synonym cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
PubChem CID 637511
ChEBI CHEBI:16731
IUPAC Name (E)-3-phenylprop-2-enal
SMILES C1=CC=C(C=C1)C=CC=O

Specifications

Assay Percent Range 99%
Quantity 100 g
Formula Weight 132.2
Concentration or Composition (by Analyte or Components) 1mL = approx. 1.05g
Chemical Name or Material Cinnamic Aldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.