missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinnamic Aldehyde, MP Biomedicals™
Click to view available options
Quantity:
1000 g
Specifications
Specifications
CAS | 104-55-2 |
Melting Point | -7.5°C (literature) |
Density | 1.0497g/cm3 at 20°C (Lit.) |
Boiling Point | 246°C (1014 hPa (literature)) |
Molecular Formula | C9H8O |
Refractive Index | n20/D 1.621 (Lit.) |
Quantity | 1000 g |
Synonym | cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal |
Solubility Information | Misc with alcohol, ether, chloroform, oils. |
InChI Key | KJPRLNWUNMBNBZ-QPJJXVBHSA-N |
Show More |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction