missing translation for 'onlineSavingsMsg'
Learn More

cis-3-(Boc-amino)cyclobutanemethanol, 97%, Thermo Scientific™

Product Code. 15429178
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15429178

Brand: Thermo Scientific Alfa Aesar H62686.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142733-64-0
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD08436074
InChI Key PCPNTJQMXAHNOA-UHFFFAOYSA-N
Synonym tert-butyl n-3-hydroxymethyl cyclobutyl carbamate, tert-butyl cis-3-hydroxymethyl cyclobutyl carbamate, 3-boc-amino-cyclobutanemethanol, tert-butyl cis-3-hydroxymethylcyclobutylcarbamate, tert-butyl trans-3-hydroxymethylcyclobutylcarbamate, tert-butyl 3-hydroxymethyl cyclobutyl carbamate, cis-1-boc-amino-3-hydroxymethyl cyclobutane, trans-1-boc-amino-3-hydroxymethyl cyclobutane, tert-butyl trans-3-hydroxymethyl cyclobutyl carbamate, cis-3-boc-amino-cyclobutanemethanol
PubChem CID 22408683
IUPAC Name tert-butyl N-[3-(hydroxymethyl)cyclobutyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CC(C1)CO

Specifications

Quantity 250 mg
Formula Weight 201.26
Percent Purity 97%
Chemical Name or Material cis-3-(Boc-amino)cyclobutanemethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.